Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Riok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100511.1 | NP_001093981.1 | 9 | 979 | ||
XM_006253789.2 | XP_006253851.1 | 9 | 1048 | |||
GenBank mRNA | BC079173.1 | - | 9 | 979 | ||
FQ209749.1 | - | 9 | 1033 | |||
FQ234746.1 | - | 6 | 445 |