Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173514.1 | NP_001166985.1 | 4 | 530 | ||
NM_001173515.1 | NP_001166986.1 | 4 | 527 | |||
NM_003332.3 | NP_003323.1 | 5 | 563 | |||
NM_198125.2 | NP_937758.1 | 5 | 560 | |||
NR_033390.1 | 4 | 523 | ||||
GenBank mRNA | AF019562.1 | - | 5 | 549 | ||
AK290385.1 | - | 5 | 546 | |||
BC011175.1 | - | 5 | 538 | 4150408 | ||
BI908458.1 | - | 4 | 493 | |||
BP295666.1 | - | 4 | 498 | |||
BP305672.1 | - | 4 | 514 |
Gene Symbol: ZNF160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X78928.1 | - | 495 |