Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106164.1 | NP_001099634.1 | 2 | 313 | ||
XM_006255049.2 | XP_006255111.1 | 3 | 396 | |||
XM_008772241.1 | XP_008770463.1 | 3 | 400 | |||
XM_008772242.1 | XP_008770464.1 | 3 | 349 | |||
GenBank mRNA | BC169107.1 | - | 3 | 405 | 7130835 |