Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106169.1 | NP_001099639.1 | 12 | 1619 | ||
XM_006255079.2 | XP_006255141.1 | 12 | 1645 | |||
XM_006255081.2 | XP_006255143.1 | 12 | 1609 | |||
XM_008772281.1 | XP_008770503.1 | 12 | 2014 |