Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011926.1 | NP_001011926.1 | 3 | 299 | ||
XM_006255181.2 | XP_006255243.1 | 3 | 311 | |||
XM_006255182.2 | XP_006255244.1 | 3 | 289 | |||
XM_006255183.2 | XP_006255245.1 | 2 | 212 | |||
XM_006255185.2 | XP_006255247.1 | 3 | 245 | |||
GenBank mRNA | BC089985.1 | - | 3 | 299 | 7318380 |