Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011926.1 | NP_001011926.1 | 9 | 919 | ||
XM_006255181.2 | XP_006255243.1 | 9 | 931 | |||
XM_006255182.2 | XP_006255244.1 | 9 | 909 | |||
XM_006255183.2 | XP_006255245.1 | 8 | 832 | |||
XM_006255185.2 | XP_006255247.1 | 9 | 865 | |||
GenBank mRNA | BC089985.1 | - | 9 | 919 | 7318380 |