Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255413.2 | XP_006255475.1 | 4 | 2138 | ||
XM_008772544.1 | XP_008770766.1 | 4 | 2138 | |||
XM_008772545.1 | XP_008770767.1 | 4 | 2138 | |||
XM_008772546.1 | XP_008770768.1 | 4 | 2138 | |||
XM_008772547.1 | XP_008770769.1 | 4 | 2138 |