Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030028.1 | NP_001025199.1 | 17 | 1900 | ||
XM_008772479.1 | XP_008770701.1 | 16 | 1932 | |||
XM_008772480.1 | XP_008770702.1 | 16 | 2004 | |||
XM_008772481.1 | XP_008770703.1 | 17 | 1851 | |||
GenBank mRNA | BC099761.1 | - | 17 | 1900 | 7930411 |