Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106201.1 | NP_001099671.1 | 5 | 2481 | ||
XM_006231634.1 | XP_006231696.1 | 6 | 3157 | |||
XM_006231635.2 | XP_006231697.1 | 6 | 3095 | |||
XM_008760523.1 | XP_008758745.1 | 6 | 3157 |