Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100518.1 | NP_001093988.1 | 6 | 990 | ||
XM_008761495.1 | XP_008759717.1 | 6 | 1046 | |||
GenBank mRNA | AY724508.1 | - | 6 | 990 | ||
BC091291.1 | - | 4 | 346 |