Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035247.2 | NP_001030324.2 | 10 | 881 | ||
NM_005659.6 | NP_005650.2 | 10 | 881 | |||
XM_011530361.2 | XP_011528663.1 | 10 | 1007 | |||
GenBank mRNA | AF141201.1 | - | 9 | 751 | ||
AJ239058.1 | - | 9 | 751 | |||
AK128120.1 | - | 5 | 2772 | |||
AK225877.1 | - | 10 | 838 | |||
AK304734.1 | - | 10 | 851 | |||
AK315998.1 | - | 10 | 832 | |||
AY101594.1 | - | 10 | 808 | |||
BC001049.2 | - | 10 | 828 | 3507963 | ||
BC005087.1 | - | 10 | 829 | 3830774 | ||
BX648483.1 | - | 10 | 813 | |||
CR456607.1 | - | 10 | 765 | |||
U64444.1 | - | 10 | 937 |