Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sft2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008302.1 | NP_001008303.1 | 4 | 360 | ||
GenBank mRNA | BC085346.1 | - | 4 | 360 | 7304630 | |
FQ225074.1 | - | 4 | 381 | |||
FQ227930.1 | - | 4 | 338 |