Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106211.1 | NP_001099681.1 | 2 | 609 | ||
XM_006229745.2 | XP_006229807.1 | 2 | 1164 |