Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001521.1 | NP_001001521.1 | 8, 9 | 1569 | ||
NM_006759.3 | NP_006750.3 | 8, 9 | 1625 | |||
XM_005264537.2 | XP_005264594.1 | 9, 10 | 1677 | |||
XM_005264538.1 | XP_005264595.1 | 9, 10 | 1644 | |||
XM_006712088.1 | XP_006712151.1 | 9, 10 | 1750 | |||
XM_011533091.1 | XP_011531393.1 | 6, 7 | 1304 | |||
XM_017004855.1 | XP_016860344.1 | 9, 10 | 1645 | |||
XM_017004856.1 | XP_016860345.1 | 10, 11 | 1537 | |||
XM_017004857.1 | XP_016860346.1 | 10, 11 | 1838 | |||
GenBank mRNA | AK300731.1 | - | 8, 9 | 1353 | ||
AK314263.1 | - | 8, 9 | 1616 | |||
BC000173.2 | - | 8, 9 | 1570 | 2985723 | ||
BC002954.1 | - | 8, 9 | 1449 | 3542849 | ||
BC047004.1 | - | 8, 9 | 1576 | 3924393 | ||
U27460.1 | - | 9, 10 | 1394 |