Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033888.1 | NP_001029060.1 | 3 | 130 | ||
XM_006228565.2 | XP_006228627.1 | 2 | 3294 | |||
XM_006228566.2 | XP_006228628.1 | 2 | 3294 | |||
XM_006228567.2 | XP_006228629.1 | 3 | 353 | |||
XM_006228568.2 | XP_006228630.1 | 3 | 559 | |||
GenBank mRNA | AB210107.1 | - | 3 | 130 | ||
BC129073.1 | - | 3 | 240 | 7189989 |