Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082549.1 | NP_001076018.1 | 5 | 733 | ||
NM_199087.1 | NP_954518.1 | 4 | 562 | |||
GenBank mRNA | BC061768.1 | - | 4 | 562 | 5621353 | |
DN934634.1 | - | 5 | 762 | |||
FQ229514.1 | - | 5 | 461 |