Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106250.1 | NP_001099720.1 | 3 | 528 | ||
XM_006228948.2 | XP_006229010.1 | 3 | 562 |