Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003363.3 | NP_003354.2 | 13, 14 | 1763 | ||
NM_199443.2 | NP_955475.1 | 12, 13 | 1622 | |||
GenBank mRNA | AF017305.1 | - | 13, 14 | 1687 | ||
AF017306.1 | - | 12, 13 | 1543 | |||
AK127055.1 | - | 7, 8 | 2199 | |||
AK222725.1 | - | 12, 13 | 1552 | |||
AK223292.1 | - | 13, 14 | 1693 | |||
AK291795.1 | - | 13, 14 | 1708 | |||
BC125130.1 | - | 13, 14 | 1734 | 40119090 | ||
BC125131.1 | - | 13, 14 | 1731 | 40119091 | ||
U20657.1 | - | 13, 14 | 1687 |