Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htatip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106263.2 | NP_001099733.1 | 1 | 70 | ||
XM_006229249.2 | XP_006229311.1 | 2 | 221 | |||
XM_006229250.2 | XP_006229312.1 | 2 | 125 | |||
XM_006229251.1 | XP_006229313.1 | 2 | 136 | |||
GenBank mRNA | BC158803.1 | - | 1 | 70 | 7321759 |