Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229414.2 | XP_006229476.1 | 3 | 545 | ||
XM_006229415.2 | XP_006229477.1 | 3 | 545 | |||
XM_008759585.1 | XP_008757807.1 | 3 | 545 | |||
XM_218828.8 | XP_218828.5 | 3 | 545 |