Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198735.2 | NP_942030.1 | 2 | 678 | ||
XM_008759694.1 | XP_008757916.1 | 2 | 906 | |||
XM_008759695.1 | XP_008757917.1 | 2 | 906 | |||
XM_008759696.1 | XP_008757918.1 | 2 | 1527 | |||
XM_008759697.1 | XP_008757919.1 | 3 | 1895 | |||
XM_008759698.1 | XP_008757920.1 | 5 | 1267 | |||
XM_008759699.1 | XP_008757921.1 | 2 | 753 | |||
XM_008759700.1 | XP_008757922.1 | 4 | 998 | |||
XM_008759701.1 | XP_008757923.1 | 2 | 821 | |||
XM_008759702.1 | XP_008757924.1 | 2 | 1527 | |||
GenBank mRNA | BC099070.1 | - | 2 | 678 | 7366134 | |
FQ227811.1 | - | 1 | 452 | |||
FQ230306.1 | - | 4 | 1014 | |||
M85193.1 | - | 2 | 655 | |||
X52082.1 | - | 2 | 648 |