Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVRAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003369.3 | NP_003360.2 | 15 | 1674 | ||
XM_011545226.2 | XP_011543528.1 | 16 | 2067 | |||
XM_017018224.1 | XP_016873713.1 | 14 | 1506 | |||
XM_017018225.1 | XP_016873714.1 | 13 | 1428 | |||
XM_017018227.1 | XP_016873716.1 | 11 | 1093 | |||
GenBank mRNA | AB012958.1 | - | 15 | 1611 | ||
AK095352.1 | - | 9 | 1251 | |||
AK222772.1 | - | 15 | 1565 | |||
AK291011.1 | - | 15 | 1652 | |||
AK293782.1 | - | 15 | 1415 | |||
AK296871.1 | - | 9 | 794 | |||
AK299978.1 | - | 3 | 411 | |||
AK316133.1 | - | 9 | 861 | |||
BC064837.1 | - | 15 | 1584 | 6186851 |