Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olr231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000206.1 | NP_001000206.1 | 1 | 496 | ||
XM_008759709.1 | XP_008757931.1 | 2 | 1205 | |||
XM_008759710.1 | XP_008757932.1 | 3 | 1715 |