- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Olr245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000215.1 | NP_001000215.1 | 1 | 824 |
Set Membership: |
G-protein coupled receptor |