Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007126.3 | NP_009057.1 | 2 | 457 | ||
GenBank mRNA | AF100752.1 | - | 2 | 280 | ||
AK307735.1 | - | 2 | 272 | |||
AK312310.1 | - | 2 | 339 | |||
BC110913.1 | - | 2 | 267 | 6502535 | ||
BC121794.1 | - | 2 | 264 | 40108360 | ||
BC122550.1 | - | 2 | 264 | 40108357 | ||
DA880782.1 | - | 2 | 457 | |||
FJ224344.1 | - | 2 | 280 | |||
FJ224352.1 | - | 2 | 280 |