Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106315.1 | NP_001099785.1 | 6 | 1449 | ||
XM_006230531.2 | XP_006230593.1 | 5 | 2617 | |||
XM_008759982.1 | XP_008758204.1 | 5 | 2617 |