Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106324.1 | NP_001099794.1 | 1123 |
Gene Symbol: LOC100911750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749036.3 | XP_003749084.1 | 1 | 1166 |