Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198738.2 | NP_942033.2 | 4 | 238 | ||
GenBank mRNA | AY239016.1 | - | 4 | 226 | ||
BC079352.1 | - | 4 | 238 | 7108218 |