Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106353.1 | NP_001099823.1 | 12 | 949 | ||
XM_006231441.2 | XP_006231503.1 | 12 | 969 | |||
GenBank mRNA | BC161938.1 | - | 12 | 901 | 9022977 | |
FQ209728.1 | - | 12 | 970 | |||
FQ221472.1 | - | 11 | 872 |