Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VRK2 SIRNA Assays ›
Gene Symbol:
VRK2
Gene Name:
vaccinia related kinase 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 57907651 - 58163991 on Build GRCh38
UniGene ID:
Hs.715298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14825
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VRK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130480.2
NP_001123952.1
6
561
NM_001130481.2
NP_001123953.1
6
403
NM_001130482.2
NP_001123954.1
6
507
NM_001130483.2
NP_001123955.1
6
561
NM_001288836.1
NP_001275765.1
7
698
NM_001288837.1
NP_001275766.1
9
1121
NM_001288838.1
NP_001275767.1
8
1003
NM_001288839.1
NP_001275768.1
5
538
NM_006296.6
NP_006287.2
6
608
XM_005264540.4
XP_005264597.1
6
621
XM_006712090.3
XP_006712153.1
6
416
XM_006712091.3
XP_006712154.1
6
656
XM_006712092.3
XP_006712155.1
7
793
XM_006712093.3
XP_006712156.1
4
498
XM_011533092.2
XP_011531394.1
6
405
XM_017004858.1
XP_016860347.1
6
561
XM_017004859.1
XP_016860348.1
7
1283
XM_017004860.1
XP_016860349.1
6
1213
XM_017004861.1
XP_016860350.1
4
312
XM_017004862.1
XP_016860351.1
5
1125
XM_017004863.1
XP_016860352.1
6
705
XM_017004864.1
XP_016860353.1
7
793
XM_017004865.1
XP_016860354.1
7
607
XM_017004866.1
XP_016860355.1
7
656
GenBank mRNA
AB000450.1
-
6
506
AJ512204.2
-
6
506
AK058199.1
-
6
463
AK223540.1
-
6
463
AK292727.1
-
8
989
AK295193.1
-
6
453
AK296611.1
-
7
683
AY228367.1
-
6
403
AY228368.1
-
6
538
AY228369.1
-
6
436
AY228370.1
-
7
628
AY228371.1
-
5
468
AY228372.1
-
6
491
BC021663.1
-
6
434
BC027854.1
-
6
559
5221315
BI906749.1
-
6
569
DB227029.1
-
6
561
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of cell shape
viral process
peptidyl-serine phosphorylation
cellular response to oxidative stress
regulation of MAPK cascade
protein autophosphorylation
regulation of interleukin-1-mediated signaling pathway
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0