Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175585.1 | NP_783175.1 | 1 | 647 |
Gene Symbol: Taar7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175586.2 | NP_783176.2 | 1 | 647 |
Gene Symbol: Taar7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009975.1 | NP_001009975.1 | 1 | 647 |
Gene Symbol: Taar7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175590.2 | NP_783180.2 | 1 | 647 |
Gene Symbol: Taar7g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175588.1 | NP_783178.1 | 1 | 647 |
Set Membership: |
![]() |