Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175590.2 | NP_783180.2 | 1 | 427 |
Gene Symbol: Taar7h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175587.1 | NP_783177.1 | 1 | 427 |
Set Membership: |
G-protein coupled receptor |