Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_003388.4 | NP_003379.3 | 13 | 2903 | ||
NM_032421.2 | NP_115797.1 | 12 | 2798 | |||
GenBank mRNA | AB006629.2 | - | 12 | 2798 | ||
AY927468.1 | - | LIT3488 AY927468 |
2 | 79 | ||
BC150332.1 | - | 12 | 2809 | 8860506 |