Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD735065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199379.2 | NP_955411.1 | 3 | 265 | ||
XM_006256172.2 | XP_006256234.1 | 2 | 680 | |||
XM_006256173.2 | XP_006256235.1 | 2 | 290 | |||
XM_006256174.2 | XP_006256236.1 | 3 | 379 | |||
XM_006256175.2 | XP_006256237.1 | 3 | 513 | |||
XM_006256176.2 | XP_006256238.1 | 4 | 474 | |||
XM_006256177.2 | XP_006256239.1 | 3 | 770 | |||
XM_006256178.2 | XP_006256240.1 | 4 | 865 | |||
XM_006256180.2 | XP_006256242.1 | 3 | 820 | |||
XM_006256181.2 | XP_006256243.1 | 2 | 678 | |||
XM_006256182.2 | XP_006256244.1 | 2 | 674 | |||
XM_008772724.1 | XP_008770946.1 | 3 | 794 | |||
XM_008772725.1 | XP_008770947.1 | 2 | 421 | |||
GenBank mRNA | BC062242.1 | - | 3 | 266 | 6888059 |