Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011945.1 | NP_001011945.1 | 14 | 1280 | ||
XM_006256283.2 | XP_006256345.1 | 15 | 1459 | |||
XM_006256285.2 | XP_006256347.1 | 12 | 1043 | |||
XM_006256286.2 | XP_006256348.1 | 15 | 1456 | |||
XM_006256287.2 | XP_006256349.1 | 14 | 1400 | |||
XM_006256288.2 | XP_006256350.1 | 15 | 1388 | |||
XM_006256289.2 | XP_006256351.1 | 15 | 1385 | |||
XM_006256290.2 | XP_006256352.1 | 13 | 2458 | |||
XM_006256291.2 | XP_006256353.1 | 14 | 1329 | |||
XM_008772874.1 | XP_008771096.1 | 13 | 2479 | |||
XM_008772875.1 | XP_008771097.1 | 13 | 1163 | |||
GenBank mRNA | BC079196.1 | - | 14 | 1280 | 7131426 |