Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WNT7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004625.3 | NP_004616.2 | 2 | 417 | ||
XM_011534091.2 | XP_011532393.1 | 3 | 164 | |||
GenBank mRNA | BC008811.2 | - | 2 | 417 | 3949579 | |
D83175.1 | - | 2 | 142 |