Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009653.1 | NP_001009653.1 | 8 | 791 | ||
GenBank mRNA | BC088473.1 | - | 8 | 791 | 7128872 |