Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013077.1 | NP_001013095.1 | 4 | 684 | ||
GenBank mRNA | BC083759.1 | - | 4 | 684 | 7192931 |