Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001377.1 | NP_001001377.1 | 1 | 296 |
Gene Symbol: LOC103691842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762001.1 | XP_008760223.1 | 1 | 296 |
Gene Symbol: Olr545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001053.1 | NP_001001053.1 | 281 |