Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003405.3 | NP_003396.1 | 2 | 878 | ||
GenBank mRNA | AK307750.1 | - | 3 | 1002 | ||
AK312650.1 | - | 2 | 722 | |||
AY007132.1 | - | 1 | 465 | |||
BC003047.1 | - | 2 | 813 | 3543571 | ||
CR456612.1 | - | 2 | 681 | |||
L20422.1 | - | 2 | 646 | |||
S80794.1 | - | 2 | 828 | |||
X57345.1 | - | 1 | 561 | |||
X78138.1 | - | 2 | 837 |