Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000042.2 | NP_000033.2 | 3, 4 | 400 | ||
GenBank mRNA | AK298416.1 | - | 3, 4 | 372 | ||
AK313838.1 | - | 3, 4 | 356 | |||
BC020703.1 | - | 3, 4 | 358 | 4767966 | ||
BC026283.1 | - | 3, 4 | 372 | 4734357 | ||
BG566874.1 | - | 3, 4 | 400 | |||
HM212425.1 | - | 2, 3 | 284 | |||
M62839.1 | - | 3, 4 | 380 | |||
S80305.1 | - | 3, 4 | 350 | |||
X53595.1 | - | 3, 4 | 374 | |||
X57847.1 | - | 3, 4 | 374 | |||
X58100.1 | - | 3, 4 | 362 |