Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168523.1 | NP_001161995.1 | 8, 9 | 872 | ||
GenBank mRNA | FQ220907.1 | - | 8, 9 | 845 | ||
FQ222771.1 | - | 8, 9 | 864 | |||
FQ229546.1 | - | 8, 9 | 866 | |||
FQ229811.1 | - | 8, 9 | 866 |