Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000377.1 | NP_001000377.1 | 396 |
Gene Symbol: LOC100909692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749541.1 | XP_003749589.1 | 1 | 396 |
Gene Symbol: LOC103691873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762131.1 | XP_008760353.1 | 1 | 108 |
Gene Symbol: Olr790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000915.1 | NP_001000915.1 | 1 | 396 |