Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000378.1 | NP_001000378.1 | 1 | 206 |
Gene Symbol: LOC502660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_578154.5 | XP_578154.3 | 1 | 239 |