Gene Symbol: APOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000042.2 | NP_000033.2 | 5, 6 | 659 | ||
GenBank mRNA | AK313838.1 | - | 5, 6 | 615 | ||
BC020703.1 | - | 5, 6 | 617 | 4767966 | ||
BC026283.1 | - | 5, 6 | 631 | 4734357 | ||
BG566874.1 | - | 5, 6 | 660 | |||
HM212425.1 | - | 4, 5 | 543 | |||
M62839.1 | - | 5, 6 | 639 | |||
S80305.1 | - | 5, 6 | 609 | |||
X53595.1 | - | 5, 6 | 633 | |||
X57847.1 | - | 5, 6 | 633 | |||
X58100.1 | - | 5, 6 | 621 |