Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFX SIRNA Assays ›
Gene Symbol:
ZFX
Gene Name:
zinc finger protein, X-linked
Gene Aliases:
ZNF926
Chromosome Location:
Chr.X: 24148994 - 24216255 on Build GRCh38
UniGene ID:
Hs.336681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14989
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178084.1
NP_001171555.1
5
949
NM_001178085.1
NP_001171556.1
8
1293
NM_001178086.1
NP_001171557.1
3
354
NM_001178095.1
NP_001171566.1
5
1028
NM_003410.3
NP_003401.2
7
1157
XM_005274591.4
XP_005274648.1
6
1323
XM_005274592.3
XP_005274649.1
5
1138
XM_005274594.4
XP_005274651.1
4
2328
XM_006724513.3
XP_006724576.1
8
1552
XM_006724515.3
XP_006724578.1
8
1554
XM_011545578.1
XP_011543880.1
8
1434
XM_011545579.1
XP_011543881.1
7
1222
XM_011545581.2
XP_011543883.1
7
1364
XM_011545583.2
XP_011543885.1
4
878
XM_017029788.1
XP_016885277.1
8
1475
XM_017029789.1
XP_016885278.1
7
2037
XM_017029790.1
XP_016885279.1
7
1442
XM_017029791.1
XP_016885280.1
6
1332
XM_017029792.1
XP_016885281.1
6
1211
XM_017029793.1
XP_016885282.1
5
1009
XM_017029794.1
XP_016885283.1
7
1263
XM_017029795.1
XP_016885284.1
4
892
XM_017029796.1
XP_016885285.1
6
1044
XM_017029797.1
XP_016885286.1
8
1503
XM_017029798.1
XP_016885287.1
4
1123
XM_017029799.1
XP_016885288.1
5
1138
XM_017029800.1
XP_016885289.1
4
1125
XM_017029801.1
XP_016885290.1
3
741
GenBank mRNA
AB209892.1
-
5
1061
AF454950.1
-
1
64
AF505617.1
-
3
238
AF505618.1
-
4
850
AY072770.1
-
1
52
BC136308.1
-
7
1145
9020295
BC136309.1
-
7
1145
9020296
M30608.1
-
6
1108
X59738.1
-
3
354
X59739.1
-
5
928
X59740.1
-
8
1277
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0