Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106518.1 | NP_001099988.1 | 11 | 1042 | ||
XM_006235119.2 | XP_006235181.1 | 11 | 1092 | |||
GenBank mRNA | BC160883.1 | - | 11 | 1023 | 7382263 | |
FQ225134.1 | - | 11 | 1023 | |||
FQ231966.1 | - | 11 | 1025 |