Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106529.1 | NP_001099999.1 | 2 | 799 | ||
XM_006235287.2 | XP_006235349.1 | 2 | 1220 | |||
XM_006235288.1 | XP_006235350.1 | 2 | 836 | |||
XM_006235289.2 | XP_006235351.1 | 2 | 1016 |