Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102417.1 | NP_001095887.1 | 642 |
Gene Symbol: LOC103690143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762549.1 | XP_008760771.1 | 2 | 945 |