Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNC2 SIRNA Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C type 2 (fast)
Gene Aliases:
-
Chromosome Location:
Chr.3: 161243747 - 161244831 on Build Rnor_6.0
UniGene ID:
Rn.43529
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s150098
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037351.1
NP_001032428.1
3
256
GenBank mRNA
DQ273679.1
-
3
256
FQ215069.1
-
320
FQ216531.1
-
320
FQ216869.1
-
320
FQ216875.1
-
350
FQ216940.1
-
312
FQ217047.1
-
320
FQ217054.1
-
320
FQ217106.1
-
349
FQ217137.1
-
320
FQ217169.1
-
320
FQ217172.1
-
320
FQ217195.1
-
320
FQ217236.1
-
331
FQ217299.1
-
320
FQ217371.1
-
320
FQ217378.1
-
320
FQ217706.1
-
320
FQ217788.1
-
320
FQ217840.1
-
320
FQ217842.1
-
320
FQ217896.1
-
319
FQ217902.1
-
320
FQ217974.1
-
349
FQ217984.1
-
320
FQ218010.1
-
320
FQ218048.1
-
350
FQ218061.1
-
320
FQ223543.1
-
320
FQ223572.1
-
320
FQ223583.1
-
320
FQ223607.1
-
320
FQ223623.1
-
320
FQ223636.1
-
320
FQ223643.1
-
320
FQ223690.1
-
320
FQ223692.1
-
320
FQ223711.1
-
320
FQ223713.1
-
320
FQ223719.1
-
320
FQ223773.1
-
332
FQ223816.1
-
341
FQ223825.1
-
320
FQ223832.1
-
320
FQ223835.1
-
350
FQ223976.1
-
320
FQ223986.1
-
320
FQ224079.1
-
320
FQ224114.1
-
349
FQ224128.1
-
320
FQ224223.1
-
320
FQ224258.1
-
320
FQ224340.1
-
320
FQ224376.1
-
335
FQ224378.1
-
320
FQ224387.1
-
320
FQ224450.1
-
320
FQ224462.1
-
320
FQ224599.1
-
320
FQ224648.1
-
320
FQ224672.1
-
320
FQ224735.1
-
321
FQ224749.1
-
320
FQ224755.1
-
349
FQ224795.1
-
320
FQ224811.1
-
320
FQ224822.1
-
320
FQ224827.1
-
317
J00793.1
-
1
40
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009394, BC024390, AK133548, CB566378.
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
actin binding
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0